1x1 image pixel for data collection
Triple Layer Hot Mill Gloves - Band Top
Triple Layer Hot Mill Gloves - Band Top
Actual product may vary
Triple Layer Hot Mill Gloves - Band Top
Ideal for forging and other operations that require heat protection.
Have Questions?
Catalog number FIS19-013544
Price (SGD)
104.00
DZN
Estimated availability date 22-Jul-2025
Add to cart
Price (SGD)
104.00
DZN
Add to cart
  • Constructed from three layers of cotton fabric
  • Burlap lining creates air pockets for increased heat protection
  • Ideal for forging and other operations that require heat protection
Specifications
ColorNatural
MaterialCotton
Product TypeHot Mill Gloves
SizeUniversal
For Use With (Application)Forging, and Hot Metal Handling/Castings/Molds
GenderMen's
Glove Cuff StyleBand Top
Liner MaterialBurlap
Series94-928
Yarn Weight28 oz.
Unit SizeDZN
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection