Gray Universal Absorbent Mats
Actual product may vary
New Pig
Gray Universal Absorbent Mats
Tough mat lasts longer and requires less changeouts
Have Questions?
Catalog NumberCapacity (English)Dimensions (L x W)Type
FIS19-003-378B28.16 oz.20 x 15 in.Universal Absorbent Pad Dispenser
Catalog number FIS19-003-378B
Price (SGD)
353.00
Pack
Capacity (English):
28.16 oz.
Dimensions (L x W):
20 x 15 in.
Type:
Universal Absorbent Pad Dispenser
Price (SGD)
353.00
Pack
Tough mat lasts longer and requires less changeouts
  • Dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind
  • Absorbed leaks and drips blend right in; dark gray color hides grime so mat stays on the job longer
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job
  • Can be wrung out and incinerated after use to reduce waste or for fuels blending
  • Dimensions: 15 x 20 in. (38.1cm x 50.8cm)
Specifications
Certifications/Compliance29 CFR 1910.22(a)(2)
ColorGray
Capacity (English)28.16 oz.
Capacity (Metric)0.83 L
MaterialPolypropylene
TypeUniversal Absorbent Pad Dispenser
For Use With (Application)Absorbs oils, coolants, solvents and water
Weight CapacityHeavyweight
Dimensions (L x W)20 x 15 in.
Length (English)20 in.
Length (Metric)50.8 cm
Width (English)15 in.
Width (Metric)38 cm
Weight (English)17 lb.
Weight (Metric)7.7 kg
Unit SizePack