▸
Facility Safety and Maintenance
▸
Spill Control and Containment
▸
Sorbents and Adsorbents
▸
Absorbent Mat Pads in Dispenser Box, Heavyweight
Actual product may vary
New Pig
Absorbent Mat Pads in Dispenser Box, Heavyweight
PIG Mat lasts 2 times longer than ordinary mats for fewer change-outs. Absorbs everything. Works everywhere.
Have Questions?
Catalog Number
Dimensions (L x W)
FIS19-003-310
13 x 10 in.
Catalog number
FIS19-003-310
Price (SGD)
234.00
Case
Add to cart
Dimensions (L x W):
13 x 10 in.
Price (SGD)
234.00
Case
Add to cart
Eight layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market; won't rip, tear or fray even when saturated
Absorbs most common industrial liquids - oils, water, solvents, coolants and more
Easy-tear perforations let you take only what you need so you use less mat and save money
Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
Heavy-weight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups
Specifications
Color
Gray
Material
Polypropylene
Type
Absorbent Mat Pads
Packaging Quantity
100 Pads
Dimensions (L x W)
13 x 10 in.
Length (English)
13 in.
Length (Metric)
33.02 cm
Width (English)
10 in.
Width (Metric)
25.4 cm