HMIG Write-On Labels
Actual product may vary
HMIG Write-On Labels
Ideal for adding vital hazard information to surfaces of small bottles, cans, and other containers.
Have Questions?
Catalog NumberLength (English)
FIS18-999-7883.88 in.
Catalog number FIS18-999-788
Price (SGD)
68.00
Pack
Length (English):
3.88 in.
Price (SGD)
68.00
Pack
Ideal for adding vital hazard information to surfaces of small bottles, cans, and other containers.
  • Hazardous Materials Identification Guide (HMIG) write on labels
  • HMIG paper labels are large enough to include the chemical name, common name and manufacturer's name
Specifications
ColorAssorted
FormatLabel
Product TypeHMIG Write-On™ Label
For Use With (Application)Ideal for adding vital hazard information to surfaces of small bottles, cans and other containers
Quantity1000/Roll
Dimensions (L x W)3.87 x 1.5 in.
Length (English)3.88 in.
Length (Metric)9.8 cm
Width (English)1.5 in.
Unit SizePack