Drum-Containment Pallets and Platforms
Actual product may vary
Eagle
Drum-Containment Pallets and Platforms
Grating removes easily for cleaning
Have Questions?
Catalog NumberTypeHeight (English)Width (English)
FIS18-999-44704 Drum Pallet, with Drain8 in.51.5 in.
Catalog number FIS18-999-4470
Price (SGD)
1,625.00
Each
Type:
4 Drum Pallet, with Drain
Height (English):
8 in.
Width (English):
51.5 in.
Price (SGD)
1,625.00
Each
Pallets and platforms contain spills and overflows of hazardous materials, keeping floors contamination-free.
  • Solid polyethylene construction provides excellent chemical resistance
  • Consist of translucent tub in safety yellow, and black grating with 1.75 in. (4.4 cm) square openings
  • Pallet units only are forkliftable
  • Poly ramp can be used with 4-drum inline platform, and with 6-drum and 8-drum low-profile platforms
Specifications
Certifications/ComplianceEPA, 40 CFR 264.175
ColorYellow
Capacity (English)66 gal.
Capacity (Metric)250 L
MaterialHDPE
Type4 Drum Pallet, with Drain
For Use With (Application)Contain spills and overflows of hazardous materials, keeping floors contamination
Load Bearing Capacity (English)8000 lb.
Load Bearing Capacity (Metric)3629 kg
ShapeSquare
Width (English)51.5 in.
Width (Metric)131 cm
Height (English)8 in.
Height (Metric)20.32 cm
Depth (English)51.5 in.
Depth (Metric)131 cm
Weight (English)67 lb.
Weight (Metric)30 kg
Unit SizeEach