Step-On Biohazard Waste Containers
Step-On Biohazard Waste Containers
Actual product may vary
Eagle
Step-On Biohazard Waste Containers
Provide safe, temporary collection of biological waste (no sharps). Foot operated pedal with self-closing lid reduces the risk of contamination. Alert workers to potential biohazards and meet OSHA requirements for exposures to bloodborne pathogens.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Height (Metric)Length (Metric)Width (Metric)
FIS18-880C14 gal.53 cm56 cm45.7 cm
FIS18-880A6 gal.41 cm42 cm34 cm
Catalog number FIS18-880C
Price (SGD)
430.00
EA
Add to cart
Capacity (English):
14 gal.
Height (Metric):
53 cm
Length (Metric):
56 cm
Width (Metric):
45.7 cm
Price (SGD)
430.00
EA
Add to cart
  • Suitable for medical facilities and research labs
  • Safely collect biological waste; reduce contamination
  • Foot lever opens container for hands-free operation
  • Heavy-wall, high density polyethylene construction won’t rust or dent
  • Labeled to advise workers of potential biohazards
Specifications
Capacity (Metric)53 L
Certifications/ComplianceFM, OSHA 29 CFR 1910.1030, NFPA
Diameter (Metric) Outer53 cm
Diameter (English) Outer21 in.
Capacity (English)14 gal.
ColorRed
Dimensions (L x W x H)22 x 18 x 21 in. (56 x 45.7 x 53 cm)
Height (Metric)53 cm
Length (Metric)56 cm
Width (Metric)45.7 cm
IncludesProminent biohazard warning label, foot lever
For Use With (Application)Suitable for medical facilities and research labs
Width (English)18 in.
Length (English)22 in.
Height (English)21 in.
MaterialHDPE
Product TypeBiohazard Waste Can
Unit SizeEA