Sodium Lactate Solution, FCC, 60%, Spectrum™ Chemical
Sodium Lactate Solution, FCC, 60%, Spectrum™ Chemical
Sodium Lactate Solution, FCC, 60%, Spectrum™ Chemical
Sodium Lactate Solution, FCC, 60%, Spectrum™ Chemical
Sodium Lactate Solution, FCC, 60%, Spectrum™ Chemical
Sodium Lactate Solution, FCC, 60%, Spectrum™ Chemical
S1217, 72-17-3, C3H5NaO3
Have Questions?
Catalog NumberQuantityPackaging
FIS18-606-605500 mLAmber Glass Bottle
Catalog number FIS18-606-605
Price (SGD)
-
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
CAS72-17-3
CAS Max %0.6
CAS7732-18-5
CAS Max %0.4
pH5.0 to 9.0
View more
Spectrum™ Chemical Sodium Lactate Solution, 60 Percent, FCC is used in food products as a preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.