Surface Spray Disinfectant
Actual product may vary
PolyCo
Surface Spray Disinfectant
Use B Braun™ Surface Spray Disinfectant for rapid surface disinfection with Rapid Meliseptrol. The broad-spectrum disinfectant has a 5-minute reaction time.
Have Questions?
Catalog NumberVolume (Metric)
FIS18-5675 L
Catalog number FIS18-567
Price (SGD)
62.00
Pack
Volume (Metric):
5 L
Price (SGD)
62.00
Pack
  • Rapid Meliseptrol distribution
  • Free of CFC's
  • For small surfaces and equipment
  • Colorless
Specifications
Product TypeDisinfectant
pH∼7
AppearanceColorless
Boiling Point89°C
Container TypeCanister
For Use With (Application)For small surfaces and equipment
Volume (Metric)5 L
Unit SizePack