▸
Primary Antibodies
▸
All Primary Antibodies
▸
IZUMO1 Antibody, DyLight 650, Novus Biologicals™
Actual product may vary
New Pig
IZUMO1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Have Questions?
Catalog Number
Quantity
FIS18-366-725
0.1 mL
Catalog number
FIS18-366-725
Price (SGD)
362.00
Case
Add to cart
Quantity:
0.1 mL
Price (SGD)
362.00
Case
Add to cart
IZUMO1 Polyclonal specifically detects IZUMO1 in Human samples. It is validated for ELISA.
Specifications
Antigen
IZUMO1
Applications
ELISA
Classification
Polyclonal
Conjugate
DyLight 650
Formulation
50 mM Sodium Borate
Gene Alias
FLJ61440, IZUMO, izumo sperm-egg fusion 1, izumo sperm-egg fusion protein 1, MGC34799, OBF, Oocyte binding/fusion factor, Sperm-specific protein izumo
Host Species
Rabbit
Immunogen
Produced in rabbits immunized with purified, recombinant IZUMO1 (AAH34769.1; Met1-Arg292).
Purification Method
Affinity purified
Quantity
0.1 mL
Regulatory Status
RUO
Primary or Secondary
Primary
Gene ID (Entrez)
284359
Target Species
Human
Form
Purified
Isotype
IgG
Unit Size
Case
Contents & Storage
Store at 4°C in the dark.