CleanTip™ Swabs: High-Sorbency Series
CleanTip™ Swabs: High-Sorbency Series
Actual product may vary
CleanTip™ Swabs: High-Sorbency Series
Certified to be silicone free
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) TipLength (Metric) Handle
FIS18-3577 mm135 mm
ITWTX700B13.5 mm126 mm
Catalog number FIS18-357
Price (SGD)
620.00
CS
Add to cart
Diameter (Metric) Tip:
7 mm
Length (Metric) Handle:
135 mm
Price (SGD)
620.00
CS
Add to cart
High Sorbency Series
Highly sorptive swabs for industrial applications such as cleaning with solvents such as IPA, applying lubricants and other liquids, and removing excess materials.
  • Cotton bud head available with or without an open-cell polyurethane foam sheath
  • Soft, nonabrasive foam cover protects against fiber release
  • Long handles constructed of hardwood for assured strength and rigidity
  • Constructed from the cleanest materials available and manufactured in a cleanroom using state-of-the-art processes and equipment
  • High-precision automated process guarantees consistent performance, sizing and cleanliness
  • Silicone-free Bag-Within-A-Bag™ packaging consists of a solvent-resistant, thermally sealed exterior bag that protects the inner bags containing the swabs
  • Each outer bag is individually lot-coded for easy traceability and quality control
Specifications
Construction100% cotton head, long Wooden handle
Diameter (English) Handle0.098 in.
Length (English)5.984 in.
For Use With (Application)Environmental sampling, application of lubricants and ointments, removal of lubricants and ointments, cleaning with solvents such as IPA or Acetone
Handle ColorBrown
Product TypeSwab
Diameter (Metric) Handle2.5 mm
Length (Metric)152 mm
Length (English) Handle5.31 in.
Length (Metric) Handle135 mm
Thickness (Metric)10 mm
Length (English) Tip0.66 in.
Length (Metric) Tip17 mm
Width (Metric) Tip3.2 mm
Width (English) Tip0.46 in.
Diameter (Metric) Tip7 mm
Diameter (English) Tip0.276 in.
Adhesive TypeAdhesive
Handle MaterialWood
Tip MaterialCotton
Unit SizeCS