Absorbond™ Polyester Blended Wipers
Absorbond™ Polyester Blended Wipers
Actual product may vary
Absorbond™ Polyester Blended Wipers
For ISO Class 6 to 7 (Class 1000 to 10,000) applications
Have Questions?
Catalog NumberDimensions (L x W)
FIS18-310A4 x 4 in. (10.16 x 10.16 cm)
Catalog number FIS18-310A
Price (SGD)
280.00
PK
Add to cart
Dimensions (L x W):
4 x 4 in. (10.16 x 10.16 cm)
Price (SGD)
280.00
PK
Add to cart
  • Hydroentangled 100% polyester wipers absorb five times their weight
  • Soft, durable surface prevents abrasion
  • Ideal wipers for cleaning oil or grease; have many optical applications
  • Contain no surfactants and have ultralow levels of particles and extractables
Specifications
Product TypeCleanroom Wipe
Length (English)4 in.
Length (Metric)10.2 cm
Width (English)4 in.
Width (Metric)10.16 cm
MaterialHydroentangled Polyester
Basis Weight42
Certifications/ComplianceISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Dimensions (L x W)4 x 4 in. (10.16 x 10.16 cm)
For Use With (Application)Cleaning, absorbing oil and grease
Unit SizePK