CD82/Kai-1 Antibody (C33), Allophycocyanin/Cy7, R&D Systems™
Actual product may vary
Excelta
CD82/Kai-1 Antibody (C33), Allophycocyanin/Cy7, R&D Systems™
A Novus product, part of R&D Systems. Mouse Monoclonal Antibody
Have Questions?
Catalog NumberQuantity
FIS18-100-9050.1 mL
Catalog number FIS18-100-905
Price (SGD)
166.00
Each
Quantity:
0.1 mL
Price (SGD)
166.00
Each
CD82/Kai-1 Monoclonal specifically detects CD82/Kai-1 in Human samples. It is validated for Flow Cytometry.
Specifications
AntigenCD82/Kai-1
ApplicationsFlow Cytometry
ClassificationMonoclonal
CloneC33
ConjugateAPC-Cyanine7
DilutionFlow Cytometry
FormulationPBS with 0.05% Sodium Azide
Gene AliasCD82 antigenC33, CD82 molecule, IA4C33 antigen, Inducible membrane protein R2, KAI1GR15, kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyteantigen, antigen detected by monoclonal and IA4)), Metastasis suppressor Kangai-1, R2, SAR2tetraspanin-27, ST6tspan-27, Suppressor of tumorigenicity 6 protein, Tetraspanin-27, Tspan-27, TSPAN274F9
Gene SymbolsCD82
Host SpeciesMouse
ImmunogenC91/PL (human HTLV-1+ T cell line).
Purification MethodProtein A purified
Quantity0.1 mL
Regulatory StatusRUO
Research DisciplineCancer
Primary or SecondaryPrimary
Gene ID (Entrez)3732.0
Test SpecificityThe antibody C33 reacts with CD82, a widely expressed cell surface protein of the tetraspanin family. CD82 is also found in endosome/lysosome compartments.
Target SpeciesHuman
Product TypeAntibody
IsotypeIgG2a
Product LineNovus
Unit SizeEach
Contents & Storage
Store at 4°C in the dark. Do not freeze.