Digi-Day Electronic Scoreboards
Actual product may vary
Accuform Signs
Digi-Day Electronic Scoreboards
Count up safe work days with attention-getting, lighted display scoreboard.
Have Questions?
Catalog NumberDisplayed Text
FIS17-998-278On The Job Safety Begins Here This Plant Has Worked 0202 Days Without An OSHA Recordable Injury Accidents are Avoidable
Catalog number FIS17-998-278
Price (SGD)
1,124.00
Each
Displayed Text:
On The Job Safety Begins Here This Plant Has Worked 0202 Days Without An OSHA Recordable Injury Accidents are Avoidable
Price (SGD)
1,124.00
Each
Shows safe work days by counting up electronically and automatically. If a lost time accident occurs, it's easy to set the display back to zero.
  • Innovatively designed frameless construction in durable, lightweight aluminum
  • Image goes to the edge, then wraps 2 in, deep for visual edge and unmistakable impact
  • Built-in hidden mounting system
  • Digital numbers advance one by one, every 24 hours
  • 2 1/2 in. digits stand out in bright red LED
  • Easy handheld remote allows user to set the time at which counter advances
  • Saves last number in event of power failure
Specifications
Electrical Requirements120V 60Hz
IncludesRemote control
MaterialAluminum
Quantity1 Ea.
Display TypeLED
For Use With (Application)Eliminate the hassle of manual updating
Height (English)28 in.
Width (English)20 in.
Displayed TextOn The Job Safety Begins Here This Plant Has Worked 0202 Days Without An OSHA Recordable Injury Accidents are Avoidable
Product TypeElectronic Scoreboard
Unit SizeEach