Tyvek™ IsoClean™ Sleeve
Actual product may vary
DuPont
Tyvek™ IsoClean™ Sleeve
Sterile sleeves provides a barrier to particles, microorganisms and non-hazardous light liquid splash.
Have Questions?
Catalog NumberPackagingThickness
FIS17-988-109Bulk Packaged4.40 mil
Catalog number FIS17-988-109
Price (SGD)
943.00
Case
Packaging:
Bulk Packaged
Thickness:
4.40 mil
Price (SGD)
943.00
Case
Tyvek sterile sleeves provides a barrier to particles, microorganisms and non-hazardous light liquid splash.
  • Garments are gamma sterilized to SAL of 10-6
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Elastic at both ends (bicep and wrist) for arm protection
  • 18 in. long
  • One size fits most
  • Full traceability on all sterilized apparel with Certificates of Sterility Available
Specifications
Product TypeControlled Environment Sleeve
SizeStandard
MaterialTyvek
ColorWhite
SterilitySterile
DisposableSingle-use
Closure TypeElastic Ends
Seam TypeBound
Cuff StyleElastic
ModelIC501B
Thickness4.40 mil
Certifications/ComplianceASTM F2101
Length (English)18 in.
Length (Metric)45.7 cm
PackagingBulk Packaged
Unit SizeCase