Gas Cylinder Brackets
Actual product may vary
First Safety
Gas Cylinder Brackets
All-steel construction wall mounted brackets
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsDimensions (L x W x H)Mounting
FIS17-987-155A1 Cylinder8 x 4 x 2 in.Wall
FIS17-987-155B2 Cylinders24 x 4 x 2 in.Wall
Catalog number FIS17-987-155A
Price (SGD)
113.00
Each
Holds:
1 Cylinder
Dimensions (L x W x H):
8 x 4 x 2 in.
Mounting:
Wall
Price (SGD)
113.00
Each
  • Straps rate 1138 psi and are the primary means of support, enabling cylinders to be held tight and secure against brackets
  • Support straps are 1.5 x 137cm (3.8 x 54 in.) long polypropylene with steel cinch buckle
  • Edges are protected with steel reinforced vinyl guarding to help maintain cylinders and provide extra grip
  • Steel parts are sealed with powder paint to assure long service life and chemical resistance
  • Optional Chain Sets (G-105) can be easily retrofit as a second means of support
  • Hold 10.16 to 30.48cm (4 to 12 in.) diameter cylinders
  • Constructed from 11 gauge hot rolled steel
Specifications
MaterialSteel
Height (Metric)5 cm
Height (English)2 in.
Dimensions (L x W x H)8 x 4 x 2 in.
Length (English)8 in.
Length (Metric)20.3 cm
Width (English)4 in.
Width (Metric)10.16 cm
Holds1 Cylinder
Gauge11g
IncludesWoven polypropylene straps, steel cinch style buckles
MountingWall
Product TypeBracket
Certifications/ComplianceNFPA1, CalOSHA, UFC
For Use With (Equipment)Gas cylinder
Weight (English)3 lb.
Weight (Metric)1.36 kg
Unit SizeEach