AlphaTec™ 56-910 Vinyl Closed-Back Aprons With Snap-on Ties
Actual product may vary
Ansell
AlphaTec™ 56-910 Vinyl Closed-Back Aprons With Snap-on Ties
With heat-sealed elastic cuffs
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS17-987-116CLarge
FIS17-987-116ASmall
FIS17-987-116BMedium
Catalog number FIS17-987-116C
Price (SGD)
933.00
Each
Size:
Large
Price (SGD)
933.00
Each
  • Neck ties, 15 in. long (38cm); waist ties, 29 in. long (74cm)
  • 8mil thick aprons have electronically welded vinyl binding strip that covers all edges for smoother finish
  • 6mil thick aprons have electronically welded binding strip around neck edge only; other edges are unhemmed
  • Available with snap-on nylon ties
  • Blue
Specifications
SizeLarge
MaterialVinyl
ColorBlue
Cuff StyleElastic
Thickness8 mil
IncludesSnap-on ties
Unit SizeEach