Bottle Jacket
Actual product may vary
Justrite
Bottle Jacket
Provides added protection of a polyethylene bottle without contaminating high-purity solvents
Have Questions?
Catalog NumberHolds
FIS17-985-79Bottles (4 L, 1 gal.)
Catalog number FIS17-985-79
Price (SGD)
132.00
Each
Holds:
Bottles (4 L, 1 gal.)
Price (SGD)
132.00
Each
  • Ideal for 4L and 1 gal. bottles
  • Place a bottle in the bottom half of the jacket and rotate lid for secure containment
  • Handle offers balanced pouring
  • Acts as a shock absorber to minimize breakage and spills due to bumps or falls
Specifications
HoldsBottles (4 L, 1 gal.)
Height (English)14 in.
Product TypeBottle Jacket
Height (Metric)35.5 cm
Unit SizeEach