Polypropylene Tools
Actual product may vary
Remco
Polypropylene Tools
Tough, nonsparking polypropylene, highly resistant to chemical attack
Have Questions?
Catalog NumberProduct TypeSize
FIS17-932-4ScraperLarge
Catalog number FIS17-932-4
Price (SGD)
18.00
Each
Product Type:
Scraper
Size:
Large
Price (SGD)
18.00
Each
  • Tough, nonsparking polypropylene tools are highly resistant to chemical attack and corrosion—ideal for cleanup of explosive materials and volatile liquids
  • Resilient and lightweight for easy handling
  • Withstand temperatures up to 80°C (176°F)
  • Economical enough for disposal after single use, or decontaminate and use again
  • Pan-type and two-piece green shovels
Specifications
Product TypeScraper
Color-codedYes
SterilityNon-sterile
Certifications/ComplianceFDA
ColorWhite
Dimensions (L x W)9.75 x 4 in. (24.8 x 10.16 cm)
For Use With (Application)Provide scratch-free use on metal surfaces, ideal for cleanup of explosive materials and volatile liquids
Length (English)9.75 in.
Length (Metric)24.8 cm
MaterialPolypropylene
SizeLarge
Temperature Range (English)176°F
Temperature Range (Metric)80°C
Width (English)4 in.
Width (Metric)10.16 cm
Unit SizeEach