Cryo-Apron™
Actual product may vary
Tempshield
Cryo-Apron™
Protection from splashes and contact cold in cryogenic and ultra-cold environments to -196°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Length (English)
FIS17-540B106 cm42 in.
FIS17-540C122 cm48 in.
Catalog number FIS17-540B
Price (SGD)
567.00
Each
Length (Metric):
106 cm
Length (English):
42 in.
Price (SGD)
567.00
Each
Protection from splashes and contact cold in cryogenic and ultra-cold environments to -196°C.
Protection from splashes and contact cold in cryogenic and ultra-cold environments to -196°C.
  • Waterproof, lightweight
  • Adjustable straps at neck and waist
  • All aprons are 24 in. (61cm) wide
  • Color: blue
Specifications
TypeCryogenic Apron
ColorBlue
Length (English)42 in.
Length (Metric)106 cm
Unit SizeEach