Available in a variety of tip angles.
Actual product may vary
Excelta
Available in a variety of tip angles.
Available in a variety of tip angles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
FIS17-468-543Vacuum Pickup Roto-pic 180° Straight Probe
FIS17-468-539Vacuum Pickup Roto-pic 45° Probe
FIS17-468-540Vacuum Pickup Roto-pic 60° Probe
FIS17-468-541Vacuum Pickup Roto-pic 45° short Probe
FIS17-468-542Vacuum Pickup Roto-pic 90° Probe
Catalog number FIS17-468-543
Price (SGD)
686.00
Each
Product Type:
Vacuum Pickup Roto-pic 180° Straight Probe
Price (SGD)
686.00
Each
Allows tips to be rotated by the wheel on the Roto-pic™ handle.
Specifications
Product TypeVacuum Pickup Roto-pic 180° Straight Probe
For Use With (Equipment)Roto-pic vacuum system
Unit SizeEach