Micro Spatulas, ESD Safe Aluminum Handle, Set of 5
Actual product may vary
Excelta
Micro Spatulas, ESD Safe Aluminum Handle, Set of 5
Micro Spatulas can be used to apply lubricants to small moving parts. Kit includes 5 micro spatulas of various colors.
Have Questions?
Catalog NumberLength (Metric)
FIS17-467-6956.4 cm
Catalog number FIS17-467-695
Price (SGD)
122.00
Each
Length (Metric):
6.4 cm
Price (SGD)
122.00
Each
  • High precision
  • ESD safe aluminum handle
  • Tips are made of AISI 1075 Carbon Steel
Specifications
Quantity5 Pk.
ColorYellow, Red, Green, Blue and Black (overall)
Product TypeMicro Spatula
For Use With (Application)Used to apply lubricants to small moving parts and for applying adhesives during electronic
Length (English)2.5 in.
PrecisionHigh
Includes5 Spatulas (260A-ESD, 260B-ESD, 260C-ESD, 260D-ESD and 260E-ESD)
Handle MaterialAluminum
MaterialAluminum
Handle TypeESD Safe Handle
Length (Metric)6.4 cm
ShapeRound End
Unit SizeEach