CoolGrip™ Aramid Fiber/Carbon Fiber Reinforced Gloves
Actual product may vary
Superior Glove
CoolGrip™ Aramid Fiber/Carbon Fiber Reinforced Gloves
Glove-within-a-glove design traps air for a high level of thermal protection against temperatures up to 320°C.
Have Questions?
Catalog NumberSize
FIS17-400-120Large
Catalog number FIS17-400-120
Price (SGD)
131.00
Pair
Size:
Large
Price (SGD)
131.00
Pair
  • Aramid Fiber blend is impervious to flame, heat
  • Glove-within-a-glove design traps air for protection against thermal contact
  • 100% wool knit terry liner provides additional insulation
  • Heat protection from temperatures up to 320°C (608°F)
  • Crafted with latex-free materials to limit the risk of allergic reactions
Specifications
Certifications/ComplianceASTM 1790-05
ColorGray
Cut Resistance RatingANSI A4
Latex FreeYes
MaterialAramid Fiber, Carbon Fiber
SizeLarge
Temperature (English) Max.608°F
Temperature (Metric) Max320°C
For Use With (Application)Automotive Assembly, and Steel Mills
Glove Cuff StyleKnit
Liner MaterialWool
SeriesSKX-W
Length (English) Cuff2.5 in.
Length (Metric) Cuff6.35 cm
Unit SizePair