Fendall Porta Stream™ II Eyewash Station
Actual product may vary
Fendall Porta Stream™ II Eyewash Station
Keep workers safe with the Honeywell Fendall Porta Stream™ II Eyewash Station, a totally self-contained station for areas without access to plumbed water.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
FIS17-292-1B180 oz. Eyesaline Concentrate
FIS19-811-411Water Additive
Catalog number FIS17-292-1B
Price (SGD)
-
Includes:
180 oz. Eyesaline Concentrate
Gravity-fed systems provide employees with soothing relief in emergency situations. Porta Stream models come with Eyesaline™ concentrate that, when mixed with tap water, produces an isotonic solution physiologically similar to human tears.
Porta Stream II Station (15 minutes)
  • Same standard features as the Porta Stream I eyewash, but with 16 gal. (61L) tank and lockable fill cap to reduce the risk of tampering
  • Comes with 180 oz. (5.3L) of Eyesaline concentrate/water additive, a green and white vinyl eyewash sign, and cast-iron hanging bracket for installation on any flat or vertical surface
  • Measures 191/2L x 191/2W x 26 in.H (50 x 50 x 66cm)
Specifications
Capacity (English)16 gal.
Capacity (Metric)60.5 L
Certifications/ComplianceANSI Z358.1- Meets ANSI Z358.1 15- Minute Flushing Requirement
ColorHi-viz Green
Depth (English)19 in.
Depth (Metric)48.26 cm
Flow Rate0.4 gpm
For Use With (Application)Emergency Eye Care
Height (English)24 in.
Height (Metric)60.96 cm
Includes180 oz. Eyesaline Concentrate
MaterialHigh Impact Polyethylene
MountingSingle Bracket Mounting Device or Transported with Optional Universal Cart or Universal Stand
Product TypeFendall Porta Stream II Eyewash station
Rinse Time15 min.
Unit SizeEach