Galvanized-Steel Oily Waste Safety Cans
Actual product may vary
Justrite
Galvanized-Steel Oily Waste Safety Cans
Use for safe disposal of oily waste, solvent cloths, and wiping rags. The galvanized-steel oily waste safety cans opens to no more than 60 degrees and stays closed when not in use.
Have Questions?
Catalog NumberCapacity (Metric)ColorClosure Type
FIS17-192B38 LRedFoot-operated Cover
Catalog number FIS17-192B
Price (SGD)
340.00
Each
Capacity (Metric):
38 L
Color:
Red
Closure Type:
Foot-operated Cover
Price (SGD)
340.00
Each
Raised, ventilated bottom promotes air circulation, prevents spontaneous combustion.
  • Rugged, lead-free galvanized-steel body with reinforced ribs
  • Trilingual safety label
  • Self-closing opens easily with foot treadle
Specifications
Capacity (Metric)38 L
Certifications/ComplianceFM, UL, OSHA
Closure TypeFoot-operated Cover
Diameter (Metric) Outer35.4 cm
Diameter (English) Outer13.9 in.
Capacity (English)10 gal.
ColorRed
FinishPowder Coated
Height (Metric)46.7 cm
Quantity1 Ea.
ShapeRound
Warranty10 Years
Weight (English)12 lb.
Weight (Metric)5.4 kg
IncludesReinforced ribs, trilingual safety label
For Use With (Application)For safe disposal of oily waste, solvent cloths and wiping rags
Height (English)18.25 in.
MaterialSteel
Product TypeVentilated Bottom Oily Waste Safety Can
Unit SizeEach