A5 Sterile Cleanroom Sleeve Protector
Actual product may vary
Ansell
A5 Sterile Cleanroom Sleeve Protector
Kimtech™ A5 Sterile Cleanroom Sleeve Protector with integrated thumb loop are triple bagged, vacuum packed, and suitable for ISO Class 5 or higher cleanrooms.
Have Questions?
Catalog NumberSize
FIS17-150-200Standard
Catalog number FIS17-150-200
Price (SGD)
1,236.00
Case
Size:
Standard
Price (SGD)
1,236.00
Case
  • A5 sterile cleanroom sleeve protectors have an average bacterial filtration efficiency (BFE) of 96%
  • Made of 100% polypropylene breathable SMS fabric
  • Triple-bagged and vacuum packaged
  • Suitable for ISO Class 5 or higher cleanrooms
  • Innovative cuff fold, which presents the inside of the sleeves as the package is opened, reducing the risk of contaminating the outside of the sleeve
  • Integrated thumb loop keeps the sleeve from rolling back and exposing the arm or wrist
  • Not made with natural rubber latex, silicone, or the BHT preservative
  • Designed for single-use
  • 100 Pairs (200 each) per case
  • Suitable for use in EU GMP ISO 5 Class A sterile cleanroom environments and laboratories
  • Each individually bagged pair of sleeves has a sterilization indicator as a visual that it's undergone gamma sterilization
RightCycle
Specifications
Product TypeProtective Sleeve
SizeStandard
MaterialPolypropylene
ColorWhite
SterilitySterile
DisposableSingle-use
Closure TypeHook and Loop Strap
Seam TypeStitched
Latex FreeYes
Model36077
Certifications/ComplianceType 6 PPE Category III
Certifications/Compliance (Europe)PPE Cat I, Regulation (EU) 2016/425
IncludesThumb Loop
Length (English)18 in.
Length (Metric)45.7 cm
Cleanroom ClassClass 100 (ISO 5)
Unit SizeCase