Check Order Status
Buffer, Reference Standard, pH 13.00 ± 0.01 at 25°C, Ricca Chemical
Element Search
Structure Search
Buffer, Reference Standard, pH 13.00 ± 0.01 at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS1625-16
500 mL
Clear PET-G Bottle
Catalog number
FIS1625-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Clear PET-G Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7447-40-7, 7732-18-5, 7447-40-7
Specifications
Chemical Name or Material
Buffer, Reference Standard
Name Note
pH 13.00± 0.01 at 25°C
CAS
7447-40-7
CAS Min %
97.0
CAS Max %
100.0
View more