1x1 image pixel for data collection
Collectors for HPLC
Collectors for HPLC
Actual product may vary
Collectors for HPLC
Collect several tubes into a single connector - no problem with the right collector! The tube connector is the most flexible connector of the SCAT system.
Have Questions?
Catalog NumberProduct TypeIncludes
FIS1601-323-in-1 CollectorFittings for 3 x 2.3/3.2 mm OD
Catalog number FIS1601-32
Price (SGD)
-
Product Type:
3-in-1 Collector
Includes:
Fittings for 3 x 2.3/3.2 mm OD
For a reliable and clean connection of hoses and capillaries for collecting liquids. SCAT collectors are available in versions with up to 8 connection options.The connections are either on the side or vertically, so that the tubes can be connected easily. Tubes with different diameters from 2.3 to 11.5 mm can be connected. The connectors are made of chemical-resistant PTFE, PFA or PP material.
Specifications
MaterialPFTE, PFA
For Use WithTube 2.3/3.2 mm O.D.
ShapeVertical
IncludesFittings for 3 x 2.3/3.2 mm OD
Product Type3-in-1 Collector
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link