Polygon Stir Bars
Actual product may vary
Polygon Stir Bars
Stir bars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Length (English)Diameter (English)
FIS16-800-50815 mm6 mm0.625 in.0.25 in.
FIS14-512-12520 mm8 mm0.78 in.0.3125 in.
FIS14-512-12625.4 mm9.5 mm1 in.0.375 in.
FIS14-512-12750.8 mm8 mm2 in.0.3125 in.
FIS14-512-12870 mm10 mm2.7 in.0.375 in.
FIS16-800-50710 mm6 mm0.375 in.0.25 in.
FIS16-800-51030 mm8 mm1.1875 in.0.3125 in.
FIS16-800-51260 mm7 mm2.3125 in.0.25 in.
FIS16-800-51380 mm10 mm3.125 in.0.375 in.
Catalog number FIS16-800-508
Price (SGD)
5.00
Each
Length (Metric):
15 mm
Diameter (Metric):
6 mm
Length (English):
0.625 in.
Diameter (English):
0.25 in.
Price (SGD)
5.00
Each
Stir bars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification
  • Autoclave safe
Specifications
AutoclavableAutoclavable
Certifications/ComplianceFDA, ISO 9001:2008
Diameter (English)0.25 in.
Length (English)0.625 in.
CoatingPTFE, FDA-grade, USP Class VI
Diameter (Metric)6 mm
Length (Metric)15 mm
MaterialAlnico Magnet
Quantity50
ShapePolygon
TypeMagnetic Stir Bar
Unit SizeEach