Check Order Status
Hazard Communication Labels
Actual product may vary
Hazard Communication Labels
Available with five different warning messages and color combinations.
Have Questions?
Change view
Catalog Number
Color
Display Text
FIS15964
Red/Yellow
RADIOACTIVE
FIS15960
Black/Orange, Orange
CORROSIVE
FIS15961
Black/Green, Green
CONTAMINATED
FIS15963
Black/Chrome
POISON
4 Options
Catalog number
FIS15964
Price (SGD)
22.00
PK
Add to cart
Color:
Red/Yellow
Display Text:
RADIOACTIVE
Price (SGD)
22.00
PK
Add to cart
Continuous adhesive tape
Specifies the type of hazard and contains a corresponding visual symbol to augment the warning
Specifications
Color
Red/Yellow
Format
Label
Product Type
Hazard Communication Label
Display Text
RADIOACTIVE
For Use With (Application)
Specifies the type of hazard and contains a corresponding visual symbol to augment the warning
Quantity
125/Pk.
Size (English)
0.5 x 4 in.
Size (Metric)
1.3 x 10.2 cm
Length (English)
500 in.
Length (Metric)
12.7 m