High-Low Temperature Cloth Tape
High-Low Temperature Cloth Tape
Actual product may vary
High-Low Temperature Cloth Tape
White, textured surface may be stamped or written on.
Have Questions?
Catalog NumberColor
FIS15947White
Catalog number FIS15947
Price (SGD)
84.00
EA
Add to cart
Color:
White
Price (SGD)
84.00
EA
Add to cart
White, textured surface may be stamped or written on.
  • Withstands temperatures as high as 287.8°C (550°F) and as low as -211.7°C (-384.8°F)
  • White, self-adhesive
Specifications
Adhesive TypeSelf-adhesive
ColorWhite
FormatTape
Product TypeHigh Low Temperature Tape
For Use With (Application)Textured surface may be stamped or written on
QuantityEach
Temperature Range (English)-384.8 to 550°F
Temperature Range (Metric)-211.7 to 287.8°C
Length (English)300 in.
Length (Metric)762 cm
Width (English)1 in.
Unit SizeEA