Ro 25-6981 maleate
Ro 25-6981 maleate
Actual product may vary
Ro 25-6981 maleate
Ro 25-6981 maleate
Ro 25-6981 maleate
GluN2B-selective NMDA antagonist
Have Questions?
Catalog NumberQuantity
FIS159411 mg
Catalog number FIS15941
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 mg
Potent and selective activity-dependent blocker of NMDA receptors containing the GluN2B (formally NR2B) subunit. IC50 values are 0.009 and 52 μM for cloned receptor subunit combinations GluN1C/GluN2B and GluN1C/GluN2A respectively. Displays neuroprotectant effects in vivo and in vitro.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
For Research Use Only
Specifications
Chemical Name or Material(αR,βS)-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate
CAS1312991-76-6
Assay Percent Range>98%
Recommended StorageDesiccate at Room Temperature
Quantity1 mg
SolubilitySoluble to 100mM in DMSO
Molecular FormulaC22H29NO2.C4H4O4
Formula WeightObserved MW: 455.55
Unit SizeEA