Single-Roll Tape Dispenser
Single-Roll Tape Dispenser
Actual product may vary
Single-Roll Tape Dispenser
Accommodates various tape widths, on 3 in. cores, for easy dispensing.
Have Questions?
Catalog NumberHolds
FIS15936Tapes with 3 in. Core
Catalog number FIS15936
Price (SGD)
92.00
EA
Add to cart
Holds:
Tapes with 3 in. Core
Price (SGD)
92.00
EA
Add to cart
Accommodates various tape widths, on 3 in. cores, for easy dispensing.
  • For 0.50, 0.75, or 1 in. wide (1.3, 1.9, or 2.5cm) tapes (see inset) on 3 in. (7.6cm) core
  • Metal construction
Specifications
Product TypeSingle-Roll Tape Dispenser
HoldsTapes with 3 in. Core
MaterialMetal
For Use With (Equipment)For 60 yd. roll of 0.50, 0.75, or 1 in. W
Unit SizeEA