Product Image
Dynabeads™ MX4 Mixer
Dynabeads™ MX4 Mixer
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS15910Dynabeads™ Sample Mixers
Catalog number FIS15910
Price (SGD)
5.00
Pack
For Use With (Equipment):
Dynabeads™ Sample Mixers
Price (SGD)
5.00
Pack
The Dynabeads™ MX4 Mixer combines Dynabeads™ MX Mixer Base (Cat. No. 159-02) and MPC™-S Mixing Head (Cat. No. 159-11). Designed to hold two MPC™-S sample racks (Cat. No. A13346); capacity up to 12 microcentrifuge tubes.
Dynabeads™ MX mixers provide continuous gentle motion to obtain optimal binding of Dynabeads™ with targets (e.g., cells, proteins, nucleic acids). Variable speed (5-40 rpm). Can be operated in both cold and warm conditions (4-37°C). Inter-changeable heads accommodate different tube sizes and sample volumes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Speed Range5 to 40 rpm
Voltage12 VDC
Temperature Range (Metric)4°C to 37°C
Amperage0.5 A
Product TypeMixer
Dimensions (L x W x H)6 x 6 x 10 in. (20 x 15 x 25 cm)
Height (English)10 in.
Height (Metric)25 cm
Length (English)6 in.
Length (Metric)20 cm
Width (English)6 in.
Width (Metric)15 cm
For Use With (Equipment)Dynabeads™ Sample Mixers
Unit SizePack