Check Order Status
Buffer, Reference Standard, pH 9.00 ± 0.01 at 25°C, Ricca Chemical
Element Search
Structure Search
Buffer, Reference Standard, pH 9.00 ± 0.01 at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS1590-16
500 mL
Natural Poly Bottle
Catalog number
FIS1590-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
10043-35-3, 7447-40-7, 1310-73-2, 7732-18-5
Specifications
Chemical Name or Material
Buffer, Reference Standard
Name Note
pH 9.00± 0.01 at 25°C
CAS
10043-35-3
Color
Colorless
CAS
7447-40-7
View more