Check Order Status
Buffer, Reference Standard, pH 7.00 ± 0.01 at 25°C (Color Coded Yellow), Ricca Chemical
Element Search
Structure Search
Buffer, Reference Standard, pH 7.00 ± 0.01 at 25°C (Color Coded Yellow), Ricca Chemical
Have Questions?
Change view
Quantity:
1 L
500 mL
Packaging:
Natural Poly Bottle
Catalog number
FIS1551-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7558-79-4, 7778-77-0, 7732-18-5
Specifications
Chemical Name or Material
Buffer, Reference Standard
Name Note
pH 7.00 ± 0.01 at 25 deg C (Color Coded Yellow)
CAS
7558-79-4
CAS Min %
97.0
CAS Max %
100.0
View more