Recirculating Chiller for Sonicators
Recirculating Chiller for Sonicators
Actual product may vary
Fisherbrand
Recirculating Chiller for Sonicators
Keeps samples cool during sonication
Have Questions?
Catalog number FIS15338421
Price (SGD)
-
For use with Qsonica Models Q500/Q700 Sonicators. Compact and powerful chiller easily automates the cooling process. It keeps samples cool while occupying very little bench space. The equipment recirculates cold water through the ultrasonic horn reservoir to automate temperature. Simply set the chiller to a desired temperature, from 2 to 45°C, and cold water will recirculate through the ultrasonic horn reservoir.
  • Benchtop model
  • Temperature control range: 2° to 45°C
  • Can be used with cup horns (431B1/431C1) or microplate horn (431MPX)
  • Electrical Reqts: 85 to 265VAC, 50/60Hz
  • Dimensions: 7D ™ 5W ™ 7.5 in.H (18 ™ 13 ™ 19cm)
  • Weight: 8 lb. (3.5kg)
Specifications
Product TypeRecirculating Chiller
Height (English)7.5 in.
Height (Metric)19 cm
Width (English)5 in.
Width (Metric)13 cm
For Use With (Application)Ideal for keeping samples cool during sonication
For Use With (Equipment)Cup horn or microplate horn of Q500/Q700 Sonicator™
Unit SizeEA