Check Order Status
Buffer, Reference Standard, pH 4.00 ± 0.01 at 25°C (Color Coded Red), Ricca Chemical
Element Search
Structure Search
Buffer, Reference Standard, pH 4.00 ± 0.01 at 25°C (Color Coded Red), Ricca Chemical
Have Questions?
Change view
Quantity:
1 L
500 mL
Packaging:
Natural Poly Bottle
Catalog number
FIS1501-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
877-24-7, 7732-18-5
Specifications
Chemical Name or Material
Buffer, Reference Standard
Name Note
pH 4.00 ± 0.01 at 25 deg C (Color Coded Red)
CAS
877-24-7
CAS Min %
97.0
CAS Max %
100.0
View more