1x1 image pixel for data collection
Biohazard Warning Tape
Biohazard Warning Tape
Actual product may vary
Biohazard Warning Tape
Ideal for taping down bench protectors to delineate hazardous areas.
Have Questions?
Catalog NumberColor
FIS15-965Red
Catalog number FIS15-965
Price (SGD)
35.00
EA
Add to cart
Color:
Red
Price (SGD)
35.00
EA
Add to cart
Ideal for taping down bench protectors to delineate hazardous areas.
  • Adhesive tape is printed with the word “BIOHAZARD” and the biohazard warning symbol
  • Prevents curling and helps prevent spills from seeping under protective matting
  • Can be cut into labels (250 labels per roll)
  • Red fluorescent color
Specifications
Adhesive TypeSelf-adhesive
ColorRed
FormatTape
Product TypeBiohazard Warning Tape
For Use With (Application)Ideal for taping down bench protectors to delineate hazardous areas
QuantityEach
Length (English)500 in.
Length (Metric)12.7 m
Width (English)0.5 in.
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link