Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Facility Safety and Maintenance
▸
Labels and Label Makers
▸
Label Tapes
▸
Labeling Tape
Actual product may vary
Applied Biosystems
Labeling Tape
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
Have Questions?
Change view
Catalog Number
Color
Width (English)
FIS15-951
Yellow
2 in.
FIS15-950
White
2 in.
FIS15-953
Blue
2 in.
FIS15-954
Red
2 in.
FIS15-958
White
1.5 in.
FIS15-959
Salmon
0.75 in.
6 Options
Catalog number
FIS15-951
Price (SGD)
11.00
Each
Add to cart
Color:
Yellow
Width (English):
2 in.
Price (SGD)
11.00
Each
Add to cart
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
Strong, durable and oil-, water- and acid-resistant
Withstands temperature from -23° to +121°C (-10° to +250°F)
Core diameter: 1 in.
Can be written on with pen, pencil, or permanent marker
Will stick to most surfaces including glass, plastic, and metal
Specifications
Color
Yellow
Format
Tape
Product Type
Label Tape
For Use With (Application)
Waterproof, Oil and Acid resistant
Quantity
1 Ea.
Temperature Range (English)
-10 to 250°F
Temperature Range (Metric)
-23°C to +121°C
Dimensions (L x W)
14 yd. x 2 in.
Length (English)
14 yd.
Length (Metric)
13 m
Width (English)
2 in.
Diameter (English)
1 in.
Diameter (Metric)
2.5 cm