Biohazard Labels
Actual product may vary
Biohazard Labels
Bright red fluorescent labels display the universal biohazard symbol for easy identification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWidth (English)Length (English)Length (Metric)Quantity
FIS15-9272 in.3 in.7.62 cm500/Pk.
FIS15-9285 in.5 in.12.7 cm250/Pk.
FIS159285 in.5 in.12.7 cm250/Pk.
FIS159272 in.3 in.7.62 cm500/Pk.
Catalog number FIS15-927
Price (SGD)
24.00
Pack
Width (English):
2 in.
Length (English):
3 in.
Length (Metric):
7.62 cm
Quantity:
500/Pk.
Price (SGD)
24.00
Pack
Bright red fluorescent labels display the universal biohazard symbol for easy identification.
  • Hazard communication labels ensure safety and efficiency
  • Identify equipment, containers, rooms, materials, experimental animals or combinations of these items that may contain potentially infectious agents
  • Easy peel-off labels with pressure-sensitive adhesive
Use in accordance with applicable federal and state regulations.
Specifications
Adhesive TypePressure-sensitive
ColorFluorescent Red, Black
FormatLabel
MaterialPaper
Product TypeBiohazard Warning Tape
For Use With (Application)Biohazard
Quantity500/Pk.
Dimensions (L x W)3 in. x 2 in.
Length (English)3 in.
Length (Metric)7.62 cm
Width (English)2 in.
Unit SizePack