Cryogenic Labels for Cryogenic Storage
Actual product may vary
Cryogenic Labels for Cryogenic Storage
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Comes in an easy-to-use dispenser box with 20 sheets per pack.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorQuantityFor Use With (Equipment)Size (English)
FIS15-910-DGreen1000/Roll1.5 to 2 mL tubes0.5 x 1.25 in.
FIS15-910-BBlue1000/Roll1.5 to 2 mL tubes0.5 x 1.25 in.
FIS15-910-FPurple1000/Roll1.5 to 2 mL tubes0.5 x 1.25 in.
FIS15-910-GWhite1000/Roll0.2mL PCR Tubes0.25 x 0.75 in.
FIS15-920-AWhite1000/Roll0.5 to 1.5mL tube0.375 in. dia.
FIS15-910-AWhite1000/Roll1.5 to 2 mL tubes0.5 x 1.25 in.
FIS15-920-CAssorted5000/Pk.0.5 to 1.5mL tube0.375 in. dia
FIS15-910-EYellow1000/Roll1.5 to 2 mL tubes0.5 x 1.25 in.
FIS15-910-CRed1000/Roll1.5 to 2 mL tubes0.5 x 1.25 in.
Catalog number FIS15-910-D
Price (SGD)
35.00
Pack
Color:
Green
Quantity:
1000/Roll
For Use With (Equipment):
1.5 to 2 mL tubes
Size (English):
0.5 x 1.25 in.
Price (SGD)
35.00
Pack
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Comes in an easy to use dispenser box with 20 sheets per pack.
Ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications. Suitable for use on microcentrifuge tubes, cryovials, racks, boxes and microtiter plates. Adhesive withstands temperatures from -196° to +121°C (-321° to +250°F)
  • No need to self-laminate
  • Thinner labels allow tubes to fit most storage systems
  • Available in a full range of colors and sizes
  • Guaranteed not to fall off under normal use (provided user instructions are followed)
  • Also offered in laser printer-compatible sheets (see Cat. Nos. 15-930 and 15-940 Series)
Cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications
Specifications
ColorGreen
FormatLabel
Product TypeCryogenic Label
For Use With (Application)Cryostorage, Liquid Nitrogen, refrigeration and ultra-low temperature freezer applications
For Use With (Equipment)1.5 to 2 mL tubes
Quantity1000/Roll
Size (English)0.5 x 1.25 in.
Size (Metric)1.2 x 3.1 cm
Temperature Range (English)-321 to 250°F
Temperature Range (Metric)-196°C to +121°C
Dimensions (L x W)0.5 in. x 1.25 in.
Unit SizePack