Clostebol acetate, Honeywell Fluka™
Clostebol acetate, Honeywell Fluka™
Fisherbrand
Clostebol acetate, Honeywell Fluka™
Analytical standard, reference material
Have Questions?
Catalog NumberQuantity
FIS15-901-111100 mg
Catalog number FIS15-901-111
Price (SGD)
9.46
Special offer
Ends: 15-Dec-2025
24.00
Save 14.54 (61%)
EA
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (SGD)
9.46
Special offer
Ends: 15-Dec-2025
24.00
Save 14.54 (61%)
EA
Add to cart
Specifications
PackagingGlass Bottle
CAS855-19-6
Health Hazard 2P273+P280
Health Hazard 3H413
Physical FormCrystalline
View more
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support