PPD-2 DPD Powder Pop™ Dispensers for Chlorine Testing
Actual product may vary
HF Scientific
PPD-2 DPD Powder Pop™ Dispensers for Chlorine Testing
Use with any DPD-based photometer or test kit
Have Questions?
Catalog NumberFor Use With (Application)For Use With (Equipment)Volume (Metric)
FIS15-393-71Total Chlorine TestingNon-HF, Hach or Hanna10 mL
Catalog number FIS15-393-71
Price (SGD)
-
For Use With (Application):
Total Chlorine Testing
For Use With (Equipment):
Non-HF, Hach or Hanna
Volume (Metric):
10 mL
Portable dispenser accurately delivers the correct volume of DPD reagent for Free or Total Chlorine—use with any DPD-based photometer or test kit.
  • Supplied with enough DPD reagent for 400 tests
  • To use, simply invert dispenser over sample and press button to dispense reagent; powder dissolves quickly in sample, leaving no messy residue behind
  • Available for 5 and 10mL sample sizes, with formulations for a variety of photometers and test kits
Specifications
Volume (Metric)10 mL
For Use With (Application)Total Chlorine Testing
For Use With (Equipment)Non-HF, Hach or Hanna
Quantity400 Tests
TypeChlorine Reagent Powder Pop Dispenser
Volume (English)0.33 oz.
Unit SizeEach