Bostwick Consistometer
Actual product may vary
Fisherbrand
Bostwick Consistometer
Accurately determines sample consistency
Have Questions?
Catalog NumberCertifications/Compliance
FIS15-347-50R-81294D and ASTM F1080-93
Catalog number FIS15-347-50
Price (SGD)
1,805.00
Each
Certifications/Compliance:
R-81294D and ASTM F1080-93
Price (SGD)
1,805.00
Each
Accurately determines sample consistency
  • Measures the distance a material flows under its own weight during a given time interval to determine consistency
  • Used extensively by preservers, producers of jams, jellies, and other highly viscous products such as tomato paste, ketchup, baby foods, cosmetics, lotions, and paint
  • Enables user to predetermine formulas to standardize production lots
  • Made of durable stainless steel and equipped with a level and two leveling screws
  • Spring-operated gate is held by a positive-release mechanism permitting instantaneous flow of sample
  • Trough is graduated in 0.5cm divisions to permit accurate measurement of flow
  • Engraved graduations won't wear off
  • Originally designed by E. P. Bostwick of the U.S. Department of Agriculture, Canned Fruits and Vegetable Service
Specifications
Product TypeBostwick Consistometer
Certifications/ComplianceR-81294D and ASTM F1080-93
IncludesLevel, 2 leveling screws, instructions
MaterialStainless Steel
Height (English)5.5 in.
Height (Metric)14 cm
Length (English)14 in.
Length (Metric)35 cm
Width (English)3.25 in.
Width (Metric)9 cm
Volume (Metric)75 mL
Weight (English)1.75 lb.
Weight (Metric)0.793 kg
Unit SizeEach