Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 615, eBioscience™, Invitrogen™
Actual product may vary
Fisherbrand
Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 615, eBioscience™, Invitrogen™
Rat Monoclonal Secondary Antibody
Have Questions?
Catalog NumberQuantity
FIS15-341-100100 μg
Catalog number FIS15-341-100
Price (SGD)
13,992.00
Each
Quantity:
100 μg
Price (SGD)
13,992.00
Each
Description: The monoclonal antibody M1-14D12 recognizes Mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies. Applications Reported: This M1-14D12 antibody has been reported for use in immunohistochemical staining and immunocytochemistry. Applications Tested: This M1-14D12 antibody has been tested by immunocytochemistry and immunohistochemistry after staining with a primary antibody of the mouse IgG1 isotype. This can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be titrated for optimal performance in the assay of interest. This product has not been validated for flow cytometric analysis. Filter Recommendation: When using this eFluor™ 615 antibody conjugate, we recommend a filter that will capture the 615 emission wavelength (for example, Excitation 560/55, 585LP, Emission 645/75). A standard Alexa Fluor™ 594 filter is acceptable. Excitation: 595 nm; Emission: 615 nm. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG1 Monoclonal antibody specifically detects Mouse IgG1 in Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
For Research Use Only.
Specifications
AntigenMouse IgG1
ApplicationsImmunocytochemistry, Immunohistochemistry (Paraffin)
ClassificationMonoclonal
CloneM1-14D12
Concentration0.2 mg/mL
ConjugateeFluor 615
FormulationPBS with 0.09% sodium azide; pH 7.2
Host SpeciesRat
Purification MethodAffinity chromatography
Quantity100 μg
Regulatory StatusRUO
Primary or SecondarySecondary
Target SpeciesMouse
FormLiquid
IsotypeIgG
Unit SizeEach
Contents & Storage
4° C, store in dark, DO NOT FREEZE!