MY Model PCR Prep Station Class 100 Enclosure
Actual product may vary
Mystaire
MY Model PCR Prep Station Class 100 Enclosure
Compact laminar flow enclosures from Mystaire™ reduce the risk of sample contamination while performing PCR.
Have Questions?
Catalog NumberIncludesWidth (English)Noise LevelWeight (English)
FIS15-338-365MY-PRE2 Prefilters, MY-HEPA18 Filter24 in.<45 dBA55 lb.
Catalog number FIS15-338-365
Price (SGD)
9,052.00
Each
Includes:
MY-PRE2 Prefilters, MY-HEPA18 Filter
Width (English):
24 in.
Noise Level:
<45 dBA
Weight (English):
55 lb.
Price (SGD)
9,052.00
Each
Built-in 254 nanometer shortwave lamp effectively decontaminates the work area between amplifications. Included safety interlocks prevent operator exposure to potentially harmful UV radiation.
  • Offers class 100 vertical laminar flow air with 15-minute UV light timer
  • Constructed of clear, polycarbonate with chemically impervious polypropylene work surface
  • Features a shelf stand with built-in four-position pipet holder
  • Optional electrical cord access port allows 110V or 220V AC
  • Nominal depth of 24 in. (61 cm)
Specifications
Product TypePCR Prep Station
Lighting TypeFluorescent, UV
MaterialPolycarbonate
Air Velocity60 LFM
Depth (English)23 in.
Depth (Metric)610 mm
Dimensions (D x W x H)23 x 23 x 18 in. (584 x 584 x 457 mm)
Height (English)30 in.
Height (Metric)762 mm
Height (Metric) Sash Opening254 mm
IncludesMY-PRE2 Prefilters, MY-HEPA18 Filter
Length (English)24 in.
Length (Metric)610 mm
Noise Level<45 dBA
Voltage110/220 V
Wattage200 W
Weight (English)55 lb.
Weight (Metric)25 kg
Width (English)24 in.
Width (Metric)508 mm
Unit SizeEach