Ultrasonic Cleaning Bath Accessory, Solid Insert Tray
Actual product may vary
Branson Ultrasonics
Ultrasonic Cleaning Bath Accessory, Solid Insert Tray
Solid tray for indirect cleaning method.
Have Questions?
Catalog NumberCapacity (Metric)For Use With (Equipment)
FIS15-337-20C2.7 LUltrasonic Bath, Model 2800
Catalog number FIS15-337-20C
Price (SGD)
390.00
Each
Capacity (Metric):
2.7 L
For Use With (Equipment):
Ultrasonic Bath, Model 2800
Price (SGD)
390.00
Each
  • Solid tray for ultrasonic cleaners to prevent parts from touching tank bottom, also easily cleans discarded soil from solid tray
  • Also ideal for sample preparation
Specifications
Product TypeSolid Insert Tray
Depth (English)3 in.
Capacity (Metric)2.7 L
Capacity (English)0.75 gal.
Depth (Metric)7.62 cm
For Use With (Equipment)Ultrasonic Bath, Model 2800
IncludesBuilt in handle
Length (English)7.5 in.
Length (Metric)19.05 cm
MaterialStainless Steel
Dimensions (L x W)7.5 x 3.5 in. (19.05 x 8.89 cm)
Width (English)3.5 in.
Width (Metric)8.89 cm
Unit SizeEach