Nalgene™ Tubing T-Type Connectors
Nalgene™ Tubing T-Type Connectors
Actual product may vary
Nalgene™ Tubing T-Type Connectors
Nalgene™ Tubing T-Type Connectors
Thermo Scientific
Nalgene™ Tubing T-Type Connectors
Maximize liquid flow with non-tapered hose barbed T-Type Connectors, molded of polypropylene resin, free of animal derived components.
Have Questions?
Catalog NumberFor Use With
FIS15-319BTubing I.D. 3/16 in.
Catalog number FIS15-319B
Price (SGD)
122.00
PK
Add to cart
For Use With:
Tubing I.D. 3/16 in.
Price (SGD)
122.00
PK
Add to cart
Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing T-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.
  • Autoclavable
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionT-Type Connector
AutoclavableAutoclavable
MaterialPolypropylene
Diameter (English) Outer3/16 in.
Diameter (Metric) Outer4.76 mm
Overall Length (English)1.21 in.
Overall Length (Metric)30.734 mm
For Use WithTubing I.D. 3/16 in.
Packaging Quantity12/Pk., 72/Cs.
Product TypeTubing Connector
Unit SizePK