Clay Pipe Iron-Wire Triangles
Actual product may vary
Humboldt
Clay Pipe Iron-Wire Triangles
Have Questions?
Catalog NumberLength (English)Length (Metric)
FIS15-285B2.5 in.6.35 cm
Catalog number FIS15-285B
Price (SGD)
10.00
Pack
Length (English):
2.5 in.
Length (Metric):
6.35 cm
Price (SGD)
10.00
Pack
Crucibles completely exposed to flame. They rest on three flanges of clay pipe stems covering iron wire of triangle.
Specifications
MaterialIron Wire
Length (English)2.5 in.
Length (Metric)6.35 cm
Gauge16g
IncludesFlanged pipe stem
Product TypeTriangle
Unit SizePack