Iron-Wire Triangles
Actual product may vary
Iron-Wire Triangles
For heating crucibles
Have Questions?
Catalog NumberLength (English)Length (Metric)
FIS15-280C2.5 in.6.4 cm
Catalog number FIS15-280C
Price (SGD)
-
Length (English):
2.5 in.
Length (Metric):
6.4 cm
  • With ceramic coverings
  • Available in pack of 12
Specifications
MaterialIron Wire
Length (English)2.5 in.
Length (Metric)6.4 cm
Gauge16g
IncludesPlain clay stem
Product TypeTriangle
For Use With (Equipment)Heating crucibles
Unit SizePack